Please login to your online account to display your discounted pricing

4-Bromobenzoic hydrazide 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromobenzoic hydrazide
CAS 5933-32-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7BrN2O
Linear Formula BrC6H4C(O)NHNH2
Formula Weight 215.05
MDL Number MFCD00007602
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC386460010

  SDS
thermo fisher scientific
AC386460010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC386460250

  SDS
thermo fisher scientific
AC386460250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC386460050

  SDS
thermo fisher scientific
AC386460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 215.05
Quantity 1g
Color Light beige
Melting Point 164° to 168°C
CAS Max % 100.0
Chemical Name or Material 4-Bromobenzoic hydrazide
Linear Formula BrC6H4C(O)NHNH2
MDL Number MFCD00007602
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 98%
Molecular Formula C7H7BrN2O
CAS 5933-32-4
Synonym 4-Bromobenzhydrazide
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.