Please login to your online account to display your discounted pricing

4'-Bromobenzo-18-crown-6, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  35723-0050

 View more versions of this product

Catalog No. AC357230050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 391.26
Quantity 5g
Color White
TSCA TSCA
Melting Point 79° to 83°C
CAS Max % 100.0
Chemical Name or Material 4'-Bromobenzo-18-crown-6
Assay Percent Range 98%
Molecular Formula C16H23BrO6
MDL Number MFCD00143200
CAS 75460-28-5
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle

Specifications

Chemical Name or Material 4'-Bromobenzo-18-crown-6
CAS 75460-28-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H23BrO6
Formula Weight 391.26
MDL Number MFCD00143200