Please login to your online account to display your discounted pricing

4'-Bromobenzo-18-crown-6, 98%, ACROS Organics™

$427.07 - $427.07

Chemical Identifiers

Chemical Name or Material 4'-Bromobenzo-18-crown-6
CAS 75460-28-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H23BrO6
Formula Weight 391.26
MDL Number MFCD00143200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC357230050

  SDS
thermo fisher scientific
35723-0050
5g Each for $427.07
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 391.26
Quantity 5g
Color White
TSCA TSCA
Melting Point 79° to 83°C
CAS Max % 100.0
Molecular Formula C16H23BrO6
Infrared Spectrum Authentic
MDL Number MFCD00143200
CAS Min % 97.5
CAS 75460-28-5
Chemical Name or Material 4'-Bromobenzo-18-crown-6
Assay Percent Range 98%
Packaging Glass bottle

Chelation/Complexation reagent