Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzhydrazide, 98+%

$720.23 - $720.23

Chemical Identifiers

Chemical Name or Material 4-Bromobenzhydrazide
CAS 5933-32-4
Molecular Formula C7H7BrN2O
Formula Weight 215.05
MDL Number MFCD00007602
Beilstein 777006
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1891822

  SDS
alfa aesar™
A1891822
100g Each for $720.23
Description & Specifications

Specifications

Formula Weight 215.05
Assay ≥98%
MDL Number MFCD00007602
Molecular Formula C7H7BrN2O
CAS 5933-32-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DG4449250
Quantity 100g
Melting Point 167° to 169°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 777006
Chemical Name or Material 4-Bromobenzhydrazide

Applications
4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2′-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

Solubility
Soluble in hot methanol (almost transparency). Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.