Please login to your online account to display your discounted pricing

4-Bromobenzenediazonium tetrafluoroborate, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate
CAS 673-40-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6 H4 Br N2 . B F4
Formula Weight 270.82
MDL Number MFCD00011894
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373280250

  SDS
thermo fisher scientific
AC373280250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Powder
Formula Weight 270.82
Quantity 25g
Color Light beige to pink-beige
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate
CAS 673-40-5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C6 H4 Br N2 . B F4
MDL Number MFCD00011894
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.