Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzeneboronic acid N-methyldiethanolamine ester, 98%

$74.51 - $209.34

Chemical Identifiers

Chemical Name or Material 4-Bromobenzeneboronic acid N-methyldiethanolamine ester
CAS 133468-58-3
Molecular Formula C11H15BBrNO2
Formula Weight 283.96
MDL Number MFCD02093957
Beilstein 3320262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1777503

 
alfa aesar™
L1777503
1g Each for $209.34

AAL17775MD

 
alfa aesar™
L17775MD
250mg Each for $74.51
Description & Specifications

Specifications

Formula Weight 283.96
Chemical Name or Material 4-Bromobenzeneboronic acid N-methyldiethanolamine ester
MDL Number MFCD02093957
Assay 98%
Molecular Formula C11H15BBrNO2
CAS 133468-58-3
Melting Point 146° to 149°C
Beilstein 3320262
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
4-Bromobenzeneboronic acid N-methyldiethanolamine ester enables selective reaction of the bromo substituent with organometallic (e.g. lithium) derivatives.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.