missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzamidine hydrochloride

Manufacturer:  Alfa Aesar™ H3783103

 View more versions of this product

Catalog No. AAH3783103



Specifications

Specifications

4-Bromobenzamidine hydrochloride
269°C to 272°C (decomposition)
55368-42-8
MFCD00464967
IMTHEBSPHHMJOJ-UHFFFAOYSA-N
4-bromobenzenecarboximidamide;hydrochloride
12207714
Crystalline Solid
White
1g
C7H8BrClN2
4-bromobenzamidine hydrochloride, 4-bromobenzimidamide hydrochloride, 4-bromobenzamidine hcl, 4-bromo-benzamidine hydrochloride, 4-bromobenzenecarboximidamide hydrochloride, 4-bromobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-bromo-, monohydrochloride, 4-bromobenzenecarboxamidine, chloride, benzenecarboximidamide,4-bromo, 4-bromobenzamidinehydrochloride
C1=CC(=CC=C1C(=N)N)Br.Cl
235.509
235.51
99%

Chemical Identifiers

55368-42-8
235.509
IMTHEBSPHHMJOJ-UHFFFAOYSA-N
12207714
C1=CC(=CC=C1C(=N)N)Br.Cl
C7H8BrClN2
MFCD00464967
4-bromobenzamidine hydrochloride, 4-bromobenzimidamide hydrochloride, 4-bromobenzamidine hcl, 4-bromo-benzamidine hydrochloride, 4-bromobenzenecarboximidamide hydrochloride, 4-bromobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-bromo-, monohydrochloride, 4-bromobenzenecarboxamidine, chloride, benzenecarboximidamide,4-bromo, 4-bromobenzamidinehydrochloride
4-bromobenzenecarboximidamide;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents