Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzamidine hydrochloride

Chemical Identifiers

Chemical Name or Material 4-Bromobenzamidine hydrochloride
CAS 55368-42-8
Formula Weight 235.51
MDL Number MFCD00464967
Synonym 4-Bromobenzenecarboximidamide hydrochloride; 4-Bromobenzimidamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3783103

 
alfa aesar™
H3783103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3783106

 
alfa aesar™
H3783106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 269° to 272°C (dec.)
Formula Weight 235.51
Chemical Name or Material 4-Bromobenzamidine hydrochloride
Color White
Quantity 1g
Physical Form Crystalline Solid
Assay 99%
MDL Number MFCD00464967
CAS 55368-42-8
Synonym 4-Bromobenzenecarboximidamide hydrochloride; 4-Bromobenzimidamide hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.