Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzaldehyde dimethyl acetal, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal
CAS 24856-58-4
Formula Weight 231.09
MDL Number MFCD01320499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55283AA

  SDS
alfa aesar™
H55283AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55283AC

  SDS
alfa aesar™
H55283AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55283AE

  SDS
alfa aesar™
H55283AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.09
Density 1.383g/mL
CAS 24856-58-4
Boiling Point 254°C
MDL Number MFCD01320499
Health Hazard 1 Warning
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.5320
Physical Form Liquid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5mL
Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal

Applications
4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.