Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobenzaldehyde diethyl acetal, 98%

$69.46 - $209.45

Chemical Identifiers

Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal
CAS 34421-94-8
Molecular Formula C11H15BrO2
Formula Weight 259.14
MDL Number MFCD01863514
Beilstein 2448775
Synonym 4-Bromo-alpha,alpha-diethoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2516314

  SDS
alfa aesar™
B2516314
25g Each for $69.46

AAB2516322

  SDS
alfa aesar™
B2516322
100g Each for $209.45
Description & Specifications

Specifications

Formula Weight 259.14
Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal
Assay 98%
Molecular Formula C11H15BrO2
MDL Number MFCD01863514
CAS 34421-94-8
Beilstein 2448775
Synonym 4-Bromo-alpha,alpha-diethoxytoluene
Boiling Point 99° to 101°C (1mmHg)
Quantity 25g
Flash Point 104°C (219°F)

Applications
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.