Learn More
4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2516314
Description
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
4-Bromobenzaldehyde diethyl acetal | |
1.274 | |
104°C (219°F) | |
1.512 | |
25 g | |
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal | |
CCOC(OCC)C1=CC=C(Br)C=C1 | |
259.14 | |
259.14 |
34421-94-8 | |
99°C to 101°C (1 mmHg) | |
C11H15BrO2 | |
MFCD01863514 | |
2448775 | |
BFSNEBVTOODGHZ-UHFFFAOYSA-N | |
1-bromo-4-(diethoxymethyl)benzene | |
688340 | |
98% |
Chemical Identifiers
34421-94-8 | |
259.14 | |
BFSNEBVTOODGHZ-UHFFFAOYSA-N | |
688340 | |
CCOC(OCC)C1=CC=C(Br)C=C1 |
C11H15BrO2 | |
MFCD01863514 | |
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal | |
1-bromo-4-(diethoxymethyl)benzene |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only