Learn More
4-Bromobenzaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106670250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromobenzaldehyde | |
98.5 | |
55.0°C to 58.0°C | |
108°C | |
98.5% min. (GC) | |
C7H5BrO | |
25 g | |
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
4-bromobenzaldehyde | |
70741 | |
99% |
1122-91-4 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
BrC6H4CHO | |
07,239 | |
Solubility in water: insoluble | |
C1=CC(=CC=C1C=O)Br | |
185.02 | |
185.02 | |
Crystalline Solid |
Chemical Identifiers
1122-91-4 | |
185.02 | |
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
4-bromobenzaldehyde |
C7H5BrO | |
ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
70741 | |
C1=CC(=CC=C1C=O)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 214-365-
RUO – Research Use Only