missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromoantipyrine, TCI America™

$186.39 - $522.52

Chemical Identifiers

CAS 5426-65-3
Molecular Formula C11H11BrN2O
Molecular Weight (g/mol) 267.126
MDL Number MFCD00020779
InChI Key QYWWSDVPNCBSIA-UHFFFAOYSA-N
PubChem CID 95032
IUPAC Name 4-bromo-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B05155G View Documents TCI America
B05155G
5g Each for $186.39
 
B051525G View Documents TCI America
B051525G
25g Each for $522.52
 
Specifications

Chemical Identifiers

5426-65-3
267.126
QYWWSDVPNCBSIA-UHFFFAOYSA-N
4-bromo-1,5-dimethyl-2-phenylpyrazol-3-one
C11H11BrN2O
MFCD00020779
95032
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)Br

Specifications

White-Yellow
5g
C11H11BrN2O
QYWWSDVPNCBSIA-UHFFFAOYSA-N
4-bromo-1,5-dimethyl-2-phenylpyrazol-3-one
95032
Crystal-Powder at 20°C
4-Bromoantipyrine
117°C
5426-65-3
MFCD00020779
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)Br
267.126
267.13
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No