missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromoanisole, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 106635000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Bromoanisole | |
104-92-7 | |
100.0 | |
223.0°C | |
Authentic | |
Glass bottle | |
1.5630 to 1.5650 | |
MFCD00000097 | |
06, 199 | |
10, 1400 | |
Solubility in water: immiscible | |
COC1=CC=C(C=C1)Br | |
187.04 | |
CHEBI:47257 | |
98% |
98% | |
97.5 | |
1.4900g/mL | |
94°C | |
97.5% min. (GC) | |
C7H7BrO | |
BrC6H4OCH3 | |
500mL | |
1.49 | |
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide | |
QJPJQTDYNZXKQF-UHFFFAOYSA-N | |
1-bromo-4-methoxybenzene | |
7730 | |
187.04 |
Chemical Identifiers
104-92-7 | |
187.04 | |
QJPJQTDYNZXKQF-UHFFFAOYSA-N | |
7730 | |
1-bromo-4-methoxybenzene |
C7H7BrO | |
MFCD00000097 | |
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide | |
CHEBI:47257 | |
COC1=CC=C(C=C1)Br |
Safety and Handling
EINECSNumber : 203-252-1
RTECSNumber : BZ8501000
TSCA : TSCA
RUO â Research Use Only