Learn More
4-(Bromoacetyl)benzonitrile, 97%, Thermo Scientific Chemicals
$76.70 - $219.88
Chemical Identifiers
CAS | 20099-89-2 |
---|---|
Molecular Formula | C9H6BrNO |
Molecular Weight (g/mol) | 224.057 |
MDL Number | MFCD00052931 |
InChI Key | LJANCPRIUMHGJE-UHFFFAOYSA-N |
Synonym | 4-2-bromoacetyl benzonitrile, 4-cyanophenacyl bromide, 4-bromoacetyl benzonitrile, 2-bromo-4'-cyanoacetophenone, benzonitrile, 4-bromoacetyl, p-cyanophenacyl bromide, 4-2-bromo-acetyl-benzonitrile, 4-2-bromoethanoyl benzonitrile, 4-2-bromoacetyl benzenecarbonitrile |
PubChem CID | 98654 |
IUPAC Name | 4-(2-bromoacetyl)benzonitrile |
SMILES | C1=CC(=CC=C1C#N)C(=O)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2538206
|
Thermo Scientific Chemicals
B2538206 |
5 g |
Each for $76.70
|
|
|||||
AAB2538214
|
Thermo Scientific Chemicals
B2538214 |
25 g |
Each for $219.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20099-89-2 | |
224.057 | |
LJANCPRIUMHGJE-UHFFFAOYSA-N | |
98654 | |
C1=CC(=CC=C1C#N)C(=O)CBr |
C9H6BrNO | |
MFCD00052931 | |
4-2-bromoacetyl benzonitrile, 4-cyanophenacyl bromide, 4-bromoacetyl benzonitrile, 2-bromo-4'-cyanoacetophenone, benzonitrile, 4-bromoacetyl, p-cyanophenacyl bromide, 4-2-bromo-acetyl-benzonitrile, 4-2-bromoethanoyl benzonitrile, 4-2-bromoacetyl benzenecarbonitrile | |
4-(2-bromoacetyl)benzonitrile |
Specifications
20099-89-2 | |
Irritating | |
MFCD00052931 | |
UN2923 | |
4-2-bromoacetyl benzonitrile, 4-cyanophenacyl bromide, 4-bromoacetyl benzonitrile, 2-bromo-4'-cyanoacetophenone, benzonitrile, 4-bromoacetyl, p-cyanophenacyl bromide, 4-2-bromo-acetyl-benzonitrile, 4-2-bromoethanoyl benzonitrile, 4-2-bromoacetyl benzenecarbonitrile | |
C1=CC(=CC=C1C#N)C(=O)CBr | |
224.057 | |
224.06 | |
4-(Bromoacetyl)benzonitrile |
91°C to 96°C | |
C9H6BrNO | |
5 g | |
2087648 | |
LJANCPRIUMHGJE-UHFFFAOYSA-N | |
4-(2-bromoacetyl)benzonitrile | |
98654 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Hazard Class: 6.1; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only