Learn More
4'-Bromoacetophenone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 106595000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4'-Bromoacetophenone | |
97.5 | |
255°C | |
Authentic | |
Glass bottle | |
BrC6H4COCH3 | |
500g | |
15,141 | |
Solubility in water: insoluble | |
CC(=O)C1=CC=C(Br)C=C1 | |
199.05 | |
199.05 |
99-90-1 | |
100.0 | |
>100°C | |
97.5% min. (GC) | |
C8H7BrO | |
MFCD00000105 | |
07,283 | |
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo | |
WYECURVXVYPVAT-UHFFFAOYSA-N | |
1-(4-bromophenyl)ethanone | |
7466 | |
98% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 202-799-3
RUO â Research Use Only