Learn More
4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1222222
Description
Labeled 4â-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLabeled 4â-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.
Solubility
Soluble in Chloroform. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.
Specifications
4'-Bromoacetophenone | |
49°C to 53°C | |
>110°C (230°F) | |
MFCD00000105 | |
386015 | |
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo | |
WYECURVXVYPVAT-UHFFFAOYSA-N | |
1-(4-bromophenyl)ethanone | |
7466 | |
98% |
99-90-1 | |
255°C to 256°C | |
C8H7BrO | |
100 g | |
14,1403 | |
Soluble in Chloroform. Insoluble in water. | |
CC(=O)C1=CC=C(Br)C=C1 | |
199.05 | |
199.05 |
Chemical Identifiers
99-90-1 | |
199.05 | |
WYECURVXVYPVAT-UHFFFAOYSA-N | |
7466 | |
CC(=O)C1=CC=C(Br)C=C1 |
C8H7BrO | |
MFCD00000105 | |
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo | |
1-(4-bromophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-799-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only