missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Bromoacetanilide 98.0+%, TCI America™

Manufacturer:  TCI America B0534500G

 View more versions of this product

Catalog No. B0534500G



Specifications

Specifications

4′-Bromoacetanilide
168°C
103-88-8
MFCD00000092
MSLICLMCQYQNPK-UHFFFAOYSA-N
N-(4-bromophenyl)acetamide
7683
Crystal-Powder at 20°C
Undesignated
500g
C8H8BrNO
4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
CC(=O)NC1=CC=C(C=C1)Br
214.062
214.06
≥98.0% (GC)

Chemical Identifiers

103-88-8
214.062
MSLICLMCQYQNPK-UHFFFAOYSA-N
7683
CC(=O)NC1=CC=C(C=C1)Br
C8H8BrNO
MFCD00000092
4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
N-(4-bromophenyl)acetamide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0196

RTECSNumber : AD9625000

TSCA : Yes

Documents