Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-trans-stilbene, 98%

$130.64 - $396.28

Chemical Identifiers

Chemical Name or Material 4-Bromo-trans-stilbene
CAS 13041-70-8
Molecular Formula C14H11Br
Formula Weight 259.15
MDL Number MFCD00155020
Synonym (E)-1-Bromo-4-(2-phenylethenyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3018903

  SDS
alfa aesar™
H3018903
1g Each for $130.64

AAH3018906

  SDS
alfa aesar™
H3018906
5g Each for $396.28
Description & Specifications

Specifications

Formula Weight 259.15
MDL Number MFCD00155020
Assay 98%
CAS 13041-70-8
Synonym (E)-1-Bromo-4-(2-phenylethenyl)benzene
Quantity 1g
Chemical Name or Material 4-Bromo-trans-stilbene
Melting Point 138° to 140°C
Molecular Formula C14H11Br
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.