missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-o-xylene (contains 3-Bromo-o-xylene), TCI America™

$73.33 - $594.31

Chemical Identifiers

CAS 583-71-1
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000086
InChI Key QOGHRLGTXVMRLM-UHFFFAOYSA-N
Synonym 4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
PubChem CID 68504
IUPAC Name 4-bromo-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B134625G View Documents TCI America
B134625G
25g Each for $73.33
 
B1346500G View Documents TCI America
B1346500G
500g Each for $594.31
 
Specifications

Chemical Identifiers

583-71-1
185.064
QOGHRLGTXVMRLM-UHFFFAOYSA-N
68504
CC1=C(C=C(C=C1)Br)C
C8H9Br
MFCD00000086
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
4-bromo-1,2-dimethylbenzene

Specifications

208°C
0°C
583-71-1
MFCD00000086
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
CC1=C(C=C(C=C1)Br)C
185.064
185.06
≥75.0% (GC)
Yellow
25g
C8H9Br
3082
QOGHRLGTXVMRLM-UHFFFAOYSA-N
4-bromo-1,2-dimethylbenzene
68504
Clear Liquid at 20°C
4-Bromo-o-xylene (contains 3-Bromo-o-xylene)
Safety and Handling

Safety and Handling

RTECSNumber : CY9020400

TSCA : Yes