Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-phenylbenzamide, 98%

$98.66 - $352.50

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-phenylbenzamide
CAS 6846-12-4
Molecular Formula C13H10BrNO
Formula Weight 276.13
MDL Number MFCD00624477
Synonym 4-Bromobenzanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059703

  SDS
alfa aesar™
H5059703
1g Each for $352.50

AAH50597MD

  SDS
alfa aesar™
H50597MD
250mg Each for $98.66
Description & Specifications

Specifications

Formula Weight 276.13
Chemical Name or Material 4-Bromo-N-phenylbenzamide
Quantity 1g
Assay 98%
CAS 6846-12-4
Synonym 4-Bromobenzanilide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H10BrNO
MDL Number MFCD00624477
Health Hazard 1 Warning

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.