Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N,N-dimethylbenzenesulfonamide, 97%

$105.58 - $475.64

Chemical Identifiers

Chemical Name or Material 4-Bromo-N,N-dimethylbenzenesulfonamide
CAS 707-60-8
Molecular Formula C8H10BrNO2S
Formula Weight 264.14
MDL Number MFCD00017834
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5057903

  SDS
alfa aesar™
H5057903
1g Each for $105.58

AAH5057906

  SDS
alfa aesar™
H5057906
5g Each for $475.64
Description & Specifications

Specifications

Formula Weight 264.14
Chemical Name or Material 4-Bromo-N,N-dimethylbenzenesulfonamide
Assay 97%
Molecular Formula C8H10BrNO2S
MDL Number MFCD00017834
CAS 707-60-8
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DB0700000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.