missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-N,N-dimethylaniline, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 222210250

 View more versions of this product

Catalog No. AC222210250


 

Specifications Description & Specifications

Specifications

4-Bromo-N, N-dimethylaniline
586-77-6
100.0
Plastic bottle
≥98.5%
C8H10BrN
200.08
12, 637
53° to 56°C
99%
98.5
Authentic
Crystalline Solid
99%
BrC6H4N(CH3)2
MFCD00000093
White to gray or light blue
25g

Chemical Identifiers

586-77-6
100.0
C8H10BrN
200.08
4-Bromo-N, N-dimethylaniline, 99%
98.5
99%
BrC6H4N(CH3)2
MFCD00000093
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 209-582-2

RTECSNumber : BW9300000

TSCA : TSCA