missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N,N-bis(4-methoxybenzyl)benzenesulfonamide, 95%

$64.00 - $203.55

Chemical Identifiers

CAS 192767-23-0
Molecular Formula C22H22BrNO4S
Molecular Weight (g/mol) 476.385
MDL Number MFCD27665298
InChI Key YYLDNQJUMNGLLQ-UHFFFAOYSA-N
Synonym n,n-bis 4-methoxybenzyl-4-bromobenzenesulfonamide, 4-bromo-n,n-bis 4-methoxybenzyl benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzene-1-sulfonamide, benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzenesulfonamide
PubChem CID 25138881
IUPAC Name 4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide
SMILES COC1=CC=C(C=C1)CN(CC2=CC=C(C=C2)OC)S(=O)(=O)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64637MD View Documents Alfa Aesar™
H64637MD
250mg Each for $64.00
 
AAH6463703 View Documents Alfa Aesar™
H6463703
1g Each for $203.55
 
Specifications

Chemical Identifiers

192767-23-0
476.385
YYLDNQJUMNGLLQ-UHFFFAOYSA-N
25138881
COC1=CC=C(C=C1)CN(CC2=CC=C(C=C2)OC)S(=O)(=O)C3=CC=C(C=C3)Br
C22H22BrNO4S
MFCD27665298
n,n-bis 4-methoxybenzyl-4-bromobenzenesulfonamide, 4-bromo-n,n-bis 4-methoxybenzyl benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzene-1-sulfonamide, benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzenesulfonamide
4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide

Specifications

250mg
C22H22BrNO4S
n,n-bis 4-methoxybenzyl-4-bromobenzenesulfonamide, 4-bromo-n,n-bis 4-methoxybenzyl benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzene-1-sulfonamide, benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzenesulfonamide
COC1=CC=C(C=C1)CN(CC2=CC=C(C=C2)OC)S(=O)(=O)C3=CC=C(C=C3)Br
476.385
476.38
95%
192767-23-0
MFCD27665298
YYLDNQJUMNGLLQ-UHFFFAOYSA-N
4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide
25138881
Solid
4-Bromo-N,N-bis(4-methoxybenzyl)benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning