missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5450606

 View more versions of this product

Catalog No. AAH5450606



Specifications

Specifications

4-Bromo-N-methylbenzylamine
>110°C (230°F)
Air Sensitive
5g
C8H10BrN
1-4-bromophenyl-n-methyl-methanamine
CNCC1=CC=C(C=C1)Br
200.079
200.08
1.461
1.565
231°C to 232°C
699-03-6
MFCD04507522
URFJXIULELMVHV-UHFFFAOYSA-N
1-(4-bromophenyl)-N-methylmethanamine
485400
97%

Chemical Identifiers

699-03-6
200.079
URFJXIULELMVHV-UHFFFAOYSA-N
485400
CNCC1=CC=C(C=C1)Br
C8H10BrN
MFCD04507522
1-4-bromophenyl-n-methyl-methanamine
1-(4-bromophenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

Documents