Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-methylaniline, 97%

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-methylaniline
CAS 6911-87-1
Formula Weight 186.05
MDL Number MFCD03030391
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5527306

  SDS
alfa aesar™
H5527306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5527303

  SDS
alfa aesar™
H5527303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5527314

  SDS
alfa aesar™
H5527314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 259° to 260°C
Formula Weight 186.05
Chemical Name or Material 4-Bromo-N-methylaniline
Refractive Index 1.6130
Density 1.482g/mL
Physical Form Liquid
Quantity 5g
Assay 97%
MDL Number MFCD03030391
CAS 6911-87-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.