missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-methylaniline, 97%

Manufacturer:  Alfa Aesar™ H5527303

 View more versions of this product

Catalog No. AAH5527303



Specifications

Specifications

4-Bromo-N-methylaniline
1.613
1g
C7H8BrN
4-bromanyl-n-methyl-aniline
CNC1=CC=C(C=C1)Br
186.052
186.05
97%
1.482 g/mL
259°C to 260°C
6911-87-1
MFCD03030391
AYVPVDWQZAAZCM-UHFFFAOYSA-N
4-bromo-N-methylaniline
2757052
Liquid

Chemical Identifiers

6911-87-1
186.052
AYVPVDWQZAAZCM-UHFFFAOYSA-N
2757052
CNC1=CC=C(C=C1)Br
C7H8BrN
MFCD03030391
4-bromanyl-n-methyl-aniline
4-bromo-N-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents