Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-Fmoc-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-Fmoc-D-phenylalanine
CAS 198545-76-5
Formula Weight 426.47
MDL Number MFCD00273459
Synonym Fmoc-D-Phe(4-Br)-OH; (R)-Fmoc-2-amino-3-(4-bromophenyl)propionoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6650403

 
alfa aesar™
H6650403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6650406

 
alfa aesar™
H6650406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 426.47
Physical Form Solid
Assay 95%
MDL Number MFCD00273459
CAS 198545-76-5
Synonym Fmoc-D-Phe(4-Br)-OH; (R)-Fmoc-2-amino-3-(4-bromophenyl)propionoic acid
Chemical Name or Material 4-Bromo-N-Fmoc-D-phenylalanine
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C