missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-(4-methoxybenzyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H55757MD

 View more versions of this product

Catalog No. AAH55757MD



Specifications

Specifications

4-Bromo-N-(4-methoxybenzyl)aniline
175357-73-0
MFCD00017822
LJKLYFDMLGYRCA-UHFFFAOYSA-N
4-bromo-N-[(4-methoxyphenyl)methyl]aniline
775227
97%
250mg
C14H14BrNO
4-bromo-n-4-methoxybenzyl aniline
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)Br
292.176
292.18

Chemical Identifiers

175357-73-0
292.176
LJKLYFDMLGYRCA-UHFFFAOYSA-N
775227
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)Br
C14H14BrNO
MFCD00017822
4-bromo-n-4-methoxybenzyl aniline
4-bromo-N-[(4-methoxyphenyl)methyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents