Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-(4-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-(4-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
Formula Weight 424.23
MDL Number MFCD26143359
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6333803

  SDS
alfa aesar™
H6333803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6333806

  SDS
alfa aesar™
H6333806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63338MD

  SDS
alfa aesar™
H63338MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 424.23
Chemical Name or Material 4-Bromo-N-(4-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
Physical Form Solid
Quantity 1g
Assay 97%
MDL Number MFCD26143359