Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide, 97%

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide
CAS 56661-32-6
Formula Weight 344.13
MDL Number MFCD00421306
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58046ME

 
alfa aesar™
H58046ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH58046MC

 
alfa aesar™
H58046MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 344.13
Chemical Name or Material 4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide
Physical Form Solid
Quantity 500mg
Assay 97%
MDL Number MFCD00421306
CAS 56661-32-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Notes
Store in a dry, cool conditions in a well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.