missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-m-xylene, TCI America™

$65.70 - $388.06

Chemical Identifiers

CAS 583-70-0
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000071
InChI Key YSFLQVNTBBUKEA-UHFFFAOYSA-N
Synonym 4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
PubChem CID 68503
IUPAC Name 1-bromo-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B099625G View Documents TCI America
B099625G
25g Each for $65.70
 
B0996250G View Documents TCI America
B0996250G
250g Each for $388.06
 
Specifications

Chemical Identifiers

583-70-0
185.064
YSFLQVNTBBUKEA-UHFFFAOYSA-N
68503
CC1=CC(=C(C=C1)Br)C
C8H9Br
MFCD00000071
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
1-bromo-2,4-dimethylbenzene

Specifications

208°C
25g
C8H9Br
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
CC1=CC(=C(C=C1)Br)C
185.064
185.06
≥97.0% (GC)
Yellow
583-70-0
MFCD00000071
YSFLQVNTBBUKEA-UHFFFAOYSA-N
1-bromo-2,4-dimethylbenzene
68503
Clear Liquid at 20°C
4-Bromo-m-xylene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4042

TSCA : Yes