Learn More
4-Bromo-m-xylene, 97%, ACROS Organics™
$270.97 - $270.97
Chemical Identifiers
CAS | 583-70-0 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.064 |
MDL Number | MFCD00000071 |
InChI Key | YSFLQVNTBBUKEA-UHFFFAOYSA-N |
Synonym | 1,3-dimethyl-4-bromobenzene, 1-bromo-2,4-dimethyl-benzene, 2,4-dimethylbromobenzene, 4-brom-m-xylol, 4-bromo-1,3-dimethylbenzene, 4-bromo-m-xylene, acmc-1aowv, benzene, 1-bromo-2,4-dimethyl, m-xylene, 4-bromo, pubchem3762 |
PubChem CID | 68503 |
IUPAC Name | 1-bromo-2,4-dimethylbenzene |
SMILES | CC1=CC(=C(C=C1)Br)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC294521000
|
Thermo Fisher Scientific
294521000 |
100g |
Each for $270.97
|
|
Chemical Identifiers
583-70-0 | |
185.064 | |
YSFLQVNTBBUKEA-UHFFFAOYSA-N | |
68503 | |
CC1=CC(=C(C=C1)Br)C |
C8H9Br | |
MFCD00000071 | |
1,3-dimethyl-4-bromobenzene, 1-bromo-2,4-dimethyl-benzene, 2,4-dimethylbromobenzene, 4-brom-m-xylol, 4-bromo-1,3-dimethylbenzene, 4-bromo-m-xylene, acmc-1aowv, benzene, 1-bromo-2,4-dimethyl, m-xylene, 4-bromo, pubchem3762 | |
1-bromo-2,4-dimethylbenzene |
Specifications
Authentic | |
Colorless | |
583-70-0 | |
100.0 | |
C8H9Br | |
MFCD00000071 | |
1,3-dimethyl-4-bromobenzene, 1-bromo-2,4-dimethyl-benzene, 2,4-dimethylbromobenzene, 4-brom-m-xylol, 4-bromo-1,3-dimethylbenzene, 4-bromo-m-xylene, acmc-1aowv, benzene, 1-bromo-2,4-dimethyl, m-xylene, 4-bromo, pubchem3762 | |
CC1=CC(=C(C=C1)Br)C | |
185.064 | |
185.06 | |
≥96.0% |
1.5500 to 1.5520 (20°C, 589nm) | |
100g | |
96.0 | |
97% | |
(CH3)2C6H3Br | |
05, II, 293 | |
YSFLQVNTBBUKEA-UHFFFAOYSA-N | |
1-bromo-2,4-dimethylbenzene | |
68503 | |
Liquid | |
4-Bromo-m-xylene, 97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 209-517-8
TSCA : TSCA