missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-m-xylene, 97%

$50.12 - $846.62

Chemical Identifiers

CAS 583-70-0
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000071
InChI Key YSFLQVNTBBUKEA-UHFFFAOYSA-N
Synonym 4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
PubChem CID 68503
IUPAC Name 1-bromo-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)Br)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1021222 View Documents Alfa Aesar™
A1021222
100g Each for $181.97
 
AAA1021214 View Documents Alfa Aesar™
A1021214
25g Each for $50.12
 
AAA1021236 View Documents Alfa Aesar™
A1021236
500g Each for $846.62
 
Specifications

Chemical Identifiers

583-70-0
185.064
YSFLQVNTBBUKEA-UHFFFAOYSA-N
68503
CC1=CC(=C(C=C1)Br)C
C8H9Br
MFCD00000071
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
1-bromo-2,4-dimethylbenzene

Specifications

78°C (174°F)
100g
C8H9Br
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
CC1=CC(=C(C=C1)Br)C
185.064
185.07
4-Bromo-m-xylene
212°C to 213°C
583-70-0
MFCD00000071
YSFLQVNTBBUKEA-UHFFFAOYSA-N
1-bromo-2,4-dimethylbenzene
68503
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA