missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-m-xylene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1021236

 View more versions of this product

Catalog No. AAA1021236



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-m-xylene
1.366
500 g
C8H9Br
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
CC1=CC=C(Br)C(C)=C1
185.06
185.07
583-70-0
212°C to 213°C
1.551
MFCD00000071
YSFLQVNTBBUKEA-UHFFFAOYSA-N
1-bromo-2,4-dimethylbenzene
68503
97%

Chemical Identifiers

583-70-0
185.06
YSFLQVNTBBUKEA-UHFFFAOYSA-N
68503
CC1=CC=C(Br)C(C)=C1
C8H9Br
MFCD00000071
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
1-bromo-2,4-dimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H332-H315-H319-H335

EINECSNumber : 209-517-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only