Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-L-phenylalanine, 98+%

Chemical Identifiers

Chemical Name or Material 4-Bromo-L-phenylalanine
CAS 24250-84-8
Molecular Formula C9H10BrNO2
Formula Weight 244.09
MDL Number MFCD00063062
UN Number UN2811
Synonym H-Phe(4-Br)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2723303

  SDS
alfa aesar™
H2723303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27233MD

  SDS
alfa aesar™
H27233MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.09
UN Number UN2811
Molecular Formula C9H10BrNO2
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Synonym H-Phe(4-Br)-OH
MDL Number MFCD00063062
CAS 24250-84-8
Assay ≥98%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Chemical Name or Material 4-Bromo-L-phenylalanine
Quantity 1g
Melting Point ≈265°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.