missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-DL-phenylalanine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 227790010
Specifications
4-Bromo-DL-phenylalanine | |
Glass bottle | |
262.0°C to 263.0°C | |
99% | |
98.5 | |
99% | |
BrC6H4CH2CH(NH2)CO2H | |
2-amino-3-4-bromo-phenyl-propionic acid, 2-amino-3-4-bromophenyl propanoic acid, 2-amino-3-4-bromophenyl propionic acid, 4-bromo-dl-phenylalanine, 4-bromo-phenylalanine, 4-bromophenylalanine, dl-4-br-phe-oh, h-dl-phe 4-br-oh, p-br-phenylalanine, p-bromo-dl-phenylalanine | |
C1=CC(=CC=C1CC(C(=O)O)N)Br | |
244.09 | |
244.09 | |
99% |
Authentic | |
White | |
1g | |
14091-15-7 | |
100.0 | |
C9H10BrNO2 | |
MFCD00002599 | |
PEMUHKUIQHFMTH-UHFFFAOYSA-N | |
2-amino-3-(4-bromophenyl)propanoic acid | |
85681 | |
Powder |
Chemical Identifiers
14091-15-7 | |
244.09 | |
PEMUHKUIQHFMTH-UHFFFAOYSA-N | |
85681 | |
C1=CC(=CC=C1CC(C(=O)O)N)Br |
C9H10BrNO2 | |
MFCD00002599 | |
2-amino-3-4-bromo-phenyl-propionic acid, 2-amino-3-4-bromophenyl propanoic acid, 2-amino-3-4-bromophenyl propionic acid, 4-bromo-dl-phenylalanine, 4-bromo-phenylalanine, 4-bromophenylalanine, dl-4-br-phe-oh, h-dl-phe 4-br-oh, p-br-phenylalanine, p-bromo-dl-phenylalanine | |
2-amino-3-(4-bromophenyl)propanoic acid |
Safety and Handling
EINECSNumber : 237-941-3