Please login to your online account to display your discounted pricing

4-Bromo-alpha-methylbenzylamine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromo-α-methylbenzylamine
CAS 24358-62-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10BrN
Linear Formula BrC6H4CH(CH3)NH2
Formula Weight 200.08
MDL Number MFCD00025548
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC290990010

  SDS
thermo fisher scientific
290990010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 200.08
Physical Form Liquid
Quantity 1g
Color Clear colorless
CAS Max % 100.0
CAS 24358-62-1
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 4-Bromo-α-methylbenzylamine
Molecular Formula C8H10BrN
Health Hazard 1 Danger
MDL Number MFCD00025548
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Linear Formula BrC6H4CH(CH3)NH2
Refractive Index 1.5665 to 1.5685 (20°C, 589nm)
Assay Percent Range 99%
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.