missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-9,9-diphenylfluorene, TCI America™

$221.53 - $768.48

Chemical Identifiers

CAS 713125-22-5
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
InChI Key PBWATBVKPGTOTB-UHFFFAOYSA-N
PubChem CID 71721492
IUPAC Name 4-bromo-9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=C(C4=CC=CC=C42)C(=CC=C3)Br)C5=CC=CC=C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B47761G View Documents TCI America
B47761G
1g Each for $221.53
 
B47765G View Documents TCI America
B47765G
5g Each for $768.48
 
Specifications

Chemical Identifiers

713125-22-5
397.315
71721492
C1=CC=C(C=C1)C2(C3=C(C4=CC=CC=C42)C(=CC=C3)Br)C5=CC=CC=C5
C25H17Br
PBWATBVKPGTOTB-UHFFFAOYSA-N
4-bromo-9,9-diphenylfluorene

Specifications

White-Yellow
1g
C25H17Br
C1=CC=C(C=C1)C2(C3=C(C4=CC=CC=C42)C(=CC=C3)Br)C5=CC=CC=C5
397.315
397.32
≥98.0% (GC)
215°C
713125-22-5
PBWATBVKPGTOTB-UHFFFAOYSA-N
4-bromo-9,9-diphenylfluorene
71721492
Crystal-Powder at 20°C
4-Bromo-9,9-diphenylfluorene
Safety and Handling

Safety and Handling

TSCA : No