missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-9,9-dimethylfluorene, 98%

$25.91 - $305.34

Chemical Identifiers

CAS 942615-32-9
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD23099415
InChI Key SXOUNESKHJIXNK-UHFFFAOYSA-N
Synonym 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene
PubChem CID 59182836
IUPAC Name 4-bromo-9,9-dimethylfluorene
SMILES CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64639MD View Documents Alfa Aesar™
H64639MD
250mg Each for $25.91
 
AAH6463903 View Documents Alfa Aesar™
H6463903
1g Each for $76.65
 
AAH6463906 View Documents Alfa Aesar™
H6463906
5g Each for $305.34
 
Specifications

Chemical Identifiers

942615-32-9
273.173
SXOUNESKHJIXNK-UHFFFAOYSA-N
59182836
CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C
C15H13Br
MFCD23099415
4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene
4-bromo-9,9-dimethylfluorene

Specifications

250mg
C15H13Br
22540802
SXOUNESKHJIXNK-UHFFFAOYSA-N
4-bromo-9,9-dimethylfluorene
59182836
Solid
4-Bromo-9,9-dimethylfluorene
942615-32-9
MFCD23099415
4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene
CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C
273.173
273.17
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning