Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-5-fluoro-2-nitrotoluene, 98%

$45.26 - $137.14

Chemical Identifiers

Chemical Name or Material 4-Bromo-5-fluoro-2-nitrotoluene
CAS 224185-19-7
Molecular Formula C7H5BrFNO2
Formula Weight 234.02
MDL Number MFCD04039299
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2755203

 
alfa aesar™
H2755203
1g Each for $45.26

AAH2755206

 
alfa aesar™
H2755206
5g Each for $137.14
Description & Specifications

Specifications

Formula Weight 234.02
Chemical Name or Material 4-Bromo-5-fluoro-2-nitrotoluene
Assay 98%
Molecular Formula C7H5BrFNO2
MDL Number MFCD04039299
CAS 224185-19-7
Melting Point 59° to 63°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.