Please login to your online account to display your discounted pricing

4-Bromo-5-chlorothiophene-2-sulfonyl chloride, ≥95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW02652CB

 View more versions of this product

Catalog No. SEW02652CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 295.99
Quantity 250mg
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
CAS 166964-35-8
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD00052094
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
Molecular Formula C4HBrCl2O2S2

Specifications

Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
CAS 166964-35-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C4HBrCl2O2S2
Formula Weight 295.99
MDL Number MFCD00052094
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.