missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-4′-propylbiphenyl, TCI America™

$169.59 - $507.23

Chemical Identifiers

CAS 58743-81-0
Molecular Formula C15H15Br
Molecular Weight (g/mol) 275.189
MDL Number MFCD00060105
InChI Key DPERLOMMOVDOHH-UHFFFAOYSA-N
PubChem CID 619019
IUPAC Name 1-bromo-4-(4-propylphenyl)benzene
SMILES CCCC1=CC=C(C=C1)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B130625G View Documents TCI America
B130625G
25g Each for $507.23
 
B13065G View Documents TCI America
B13065G
5g Each for $169.59
 
Specifications

Chemical Identifiers

58743-81-0
275.189
DPERLOMMOVDOHH-UHFFFAOYSA-N
1-bromo-4-(4-propylphenyl)benzene
C15H15Br
MFCD00060105
619019
CCCC1=CC=C(C=C1)C2=CC=C(C=C2)Br

Specifications

White-Yellow
25g
C15H15Br
DPERLOMMOVDOHH-UHFFFAOYSA-N
1-bromo-4-(4-propylphenyl)benzene
619019
Crystal-Powder at 20°C
4-Bromo-4′-propylbiphenyl
108°C
58743-81-0
MFCD00060105
CCCC1=CC=C(C=C1)C2=CC=C(C=C2)Br
275.189
275.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No