Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-4'-methylbiphenyl, 97%

$164.70 - $575.69

Chemical Identifiers

Chemical Name or Material 4-Bromo-4'-methylbiphenyl
CAS 50670-49-0
Molecular Formula C13H11Br
Formula Weight 247.14
MDL Number MFCD00466236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059603

  SDS
alfa aesar™
H5059603
1g Each for $575.69

AAH50596MD

  SDS
alfa aesar™
H50596MD
250mg Each for $164.70
Description & Specifications

Specifications

Formula Weight 247.14
Molecular Formula C13H11Br
Chemical Name or Material 4-Bromo-4'-methylbiphenyl
Melting Point 136°C
Quantity 1g
Assay 97%
MDL Number MFCD00466236
CAS 50670-49-0

Applications
4-Bromo-4′-methylbiphenyl is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.