Learn More
4-Bromo-4'-hydroxybiphenyl, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1081922
Description
A biphenyl starting material. 4-Bromo-(1,1-biphenyl)-4-ol is a useful intermediate. Vinylation of 4-bromo-4-hydroxybiphenyl and ethyl acrylate using Pd (OAc) 2/PPh 3 catalyst was studied. Ethyl 4-(4-hydroxyphenyl) cinnamate was formed as the vinylation product, while, 4-hydroxybiphenyl and ethyl cinnamate were formed as side products. Preparation of 4-cyano-4'-hydroxybiphenyl This was prepared from 4-bromo-4'-benzenesulphonyloxybiphenyl by first hydrolysing it to 4-bromo-4-hydroxybiphenyl using sodium hydroxide dissolved in a mixture of water and dioxan. The syntheses of the Nanocomposite dendrimers based on cyclic phosphazene cores: Amorphous materials, were accomplished by following a modified literature procedure by reacting phosphonitrilic chloride trimer with 4-bromophenol or 4-bromo-4-hydroxybiphenyl, respectively, in the presence of K 2 CO 3 in tetrahydrofuran (THF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA biphenyl starting material. 4′-Bromo-(1,1′-biphenyl)-4-ol is a useful intermediate. Vinylation of 4-bromo-4′-hydroxybiphenyl and ethyl acrylate using Pd (OAc) 2/PPh 3 catalyst was studied. Ethyl 4-(4′-hydroxyphenyl) cinnamate was formed as the vinylation product, while, 4-hydroxybiphenyl and ethyl cinnamate were formed as side products. Preparation of 4-cyano-4′-hydroxybiphenyl This was prepared from 4-bromo-4′-benzenesulphonyloxybiphenyl by first hydrolysing it to 4-bromo-4-hydroxybiphenyl using sodium hydroxide dissolved in a mixture of water and dioxan. The syntheses of the Nanocomposite dendrimers based on cyclic phosphazene cores: Amorphous materials, were accomplished by following a modified literature procedure by reacting phosphonitrilic chloride trimer with 4-bromophenol or 4-bromo-4′-hydroxybiphenyl, respectively, in the presence of K 2 CO 3 in tetrahydrofuran (THF).
Solubility
Soluble in water (partly), and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4-Bromo-4'-hydroxybiphenyl | |
163°C to 168°C | |
MFCD00059076 | |
4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025 | |
ARUBXNBYMCVENE-UHFFFAOYSA-N | |
4-(4-bromophenyl)phenol | |
95093 | |
98% |
29558-77-8 | |
C12H9BrO | |
100 g | |
Soluble in water (partly),and methanol. | |
C1=CC(=CC=C1C2=CC=C(C=C2)Br)O | |
249.107 | |
249.11 |
Chemical Identifiers
29558-77-8 | |
249.107 | |
ARUBXNBYMCVENE-UHFFFAOYSA-N | |
95093 | |
C1=CC(=CC=C1C2=CC=C(C=C2)Br)O |
C12H9BrO | |
MFCD00059076 | |
4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025 | |
4-(4-bromophenyl)phenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only