missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-4'-(diphenylamino)biphenyl 93.0+%, TCI America™
$265.44 - $729.95
Chemical Identifiers
CAS | 202831-65-0 |
---|---|
Molecular Formula | C24H18BrN |
Molecular Weight (g/mol) | 400.32 |
MDL Number | MFCD09038499 |
InChI Key | NKCKVJVKWGWKRK-UHFFFAOYSA-N |
Synonym | (4′C-Bromobiphenyl-4-yl)diphenylamine |
PubChem CID | 11315550 |
IUPAC Name | 4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine |
SMILES | BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B2844250MG
|
TCI America
B2844250MG |
250 mg |
Each for $265.44
|
|
|||||
B28441G
|
TCI America
B28441G |
1 g |
Each for $729.95
|
|
|||||
Chemical Identifiers
202831-65-0 | |
400.32 | |
NKCKVJVKWGWKRK-UHFFFAOYSA-N | |
11315550 | |
BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H18BrN | |
MFCD09038499 | |
(4′C-Bromobiphenyl-4-yl)diphenylamine | |
4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine |
Specifications
202831-65-0 | |
C24H18BrN | |
250 mg | |
NKCKVJVKWGWKRK-UHFFFAOYSA-N | |
4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine | |
11315550 | |
≥93.0% (GC) | |
4-Bromo-4′-(diphenylamino)biphenyl |
129°C | |
MFCD09038499 | |
(4′C-Bromobiphenyl-4-yl)diphenylamine | |
BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 | |
400.32 | |
400.32 | |
Crystalline Powder |
Safety and Handling
TSCA : No