missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromo-4-chlorobutyrophenone, 97%, ACROS Organics™

Chemical Identifiers

CAS 4559-96-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.54
MDL Number MFCD00001006
InChI Key WJKPUMBLABGUCQ-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-4-chloro-1-butanone, 1-4-bromophenyl-4-chlorobutan-1-one, 1-butanone, 1-4-bromophenyl-4-chloro, 4'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-chloro-p-bromobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, acmc-20amn1
PubChem CID 78317
IUPAC Name 1-(4-bromophenyl)-4-chlorobutan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC106850250
SDS View Documents
Thermo Fisher Scientific
AC106850250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC106851000
SDS View Documents
Thermo Fisher Scientific
AC106851000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4559-96-0
261.54
WJKPUMBLABGUCQ-UHFFFAOYSA-N
78317
C1=CC(=CC=C1C(=O)CCCCl)Br
C10H10BrClO
MFCD00001006
1-4-bromophenyl-4-chloro-1-butanone, 1-4-bromophenyl-4-chlorobutan-1-one, 1-butanone, 1-4-bromophenyl-4-chloro, 4'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-chloro-p-bromobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, acmc-20amn1
1-(4-bromophenyl)-4-chlorobutan-1-one

Specifications

>110°C
35°C to 38°C
4559-96-0
C10H10BrClO
MFCD00001006
Solubility in water: insoluble
C1=CC(=CC=C1C(=O)CCCCl)Br
261.54
261.54
4'-Bromo-4-chlorobutyrophenone
Glass Bottle
25g
96% min. (GC)
BrC6H4CO(CH2)3Cl
1-4-bromophenyl-4-chloro-1-butanone, 1-4-bromophenyl-4-chlorobutan-1-one, 1-butanone, 1-4-bromophenyl-4-chloro, 4'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-chloro-p-bromobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, acmc-20amn1
WJKPUMBLABGUCQ-UHFFFAOYSA-N
1-(4-bromophenyl)-4-chlorobutan-1-one
78317
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 224-930-3

TSCA : TSCA

SDS