missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-4′,4′′-dimethyltriphenylamine, TCI America™

$166.53 - $605.02

Chemical Identifiers

CAS 58047-42-0
Molecular Formula C20H18BrN
Molecular Weight (g/mol) 352.275
InChI Key YMNJJMJHTXGFOR-UHFFFAOYSA-N
Synonym 4-Bromo-N,N-di-p-tolylaniline
PubChem CID 11142701
IUPAC Name N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30891G View Documents TCI America
B30891G
1g Each for $166.53
 
B30895G View Documents TCI America
B30895G
5g Each for $605.02
 
Specifications

Chemical Identifiers

58047-42-0
352.275
4-Bromo-N,N-di-p-tolylaniline
N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
C20H18BrN
YMNJJMJHTXGFOR-UHFFFAOYSA-N
11142701
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)Br

Specifications

White-Yellow
1g
C20H18BrN
YMNJJMJHTXGFOR-UHFFFAOYSA-N
N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
11142701
Crystal-Powder at 20°C
4-Bromo-4′,4′′-dimethyltriphenylamine
102°C
58047-42-0
4-Bromo-N,N-di-p-tolylaniline
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)Br
352.275
352.28
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No