missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)phenol, 98%

$95.92 - $444.14

Chemical Identifiers

CAS 320-49-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD03095342
InChI Key VOWPIDJSINRFPZ-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl phenol, 2-bromo-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-bromophenol, phenol, 4-bromo-3-trifluoromethyl, 4-bromo-3-trifluromethylphenol, 4-bromo-3-trifluoromethyl-phenol, btfp, pubchem2111, acmc-1ahmz, 3-trifluoromethyl-4-bromo phenol
PubChem CID 22277307
IUPAC Name 4-bromo-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6458706 View Documents Alfa Aesar™
H6458706
5g Each for $95.92
 
AAH6458714 View Documents Alfa Aesar™
H6458714
25g Each for $444.14
 
Specifications

Chemical Identifiers

320-49-0
241.007
VOWPIDJSINRFPZ-UHFFFAOYSA-N
22277307
C1=CC(=C(C=C1O)C(F)(F)F)Br
C7H4BrF3O
MFCD03095342
4-bromo-3-trifluoromethyl phenol, 2-bromo-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-bromophenol, phenol, 4-bromo-3-trifluoromethyl, 4-bromo-3-trifluromethylphenol, 4-bromo-3-trifluoromethyl-phenol, btfp, pubchem2111, acmc-1ahmz, 3-trifluoromethyl-4-bromo phenol
4-bromo-3-(trifluoromethyl)phenol

Specifications

Yellow
5g
C7H4BrF3O
UN3261
VOWPIDJSINRFPZ-UHFFFAOYSA-N
4-bromo-3-(trifluoromethyl)phenol
22277307
Solid
4-Bromo-3-(trifluoromethyl)phenol
44°C to 46°C
320-49-0
MFCD03095342
4-bromo-3-trifluoromethyl phenol, 2-bromo-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-bromophenol, phenol, 4-bromo-3-trifluoromethyl, 4-bromo-3-trifluromethylphenol, 4-bromo-3-trifluoromethyl-phenol, btfp, pubchem2111, acmc-1ahmz, 3-trifluoromethyl-4-bromo phenol
C1=CC(=C(C=C1O)C(F)(F)F)Br
241.007
241.01
98%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.