Please login to your online account to display your discounted pricing

4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C7H3BrClF3O2S
Formula Weight 323.52
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442380010

 
thermo fisher scientific
AC442380010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442380050

 
thermo fisher scientific
AC442380050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 323.52
Quantity 1g
Assay Percent Range 97%
Molecular Formula C7H3BrClF3O2S
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride
CAS 351003-47-9